Learn More
Medchemexpress LLC HY-100795A 5mg Medchemexpress, Pirmenol (hydrochloride) CAS:61477-94-9 Purity:>98%
SDP
Catalog No. 502080602
Encompass_Preferred
Medchemexpress, HY-100795A 5mg Pirmenol (hydrochloride) CAS:61477-94-9 Pirmenol hydrochloride inhibits IK.ACh by blocking muscarinic receptors. The IC50 of Pirmenol for inhibition of Carbachol-induced IK.ACh is 0.1 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0602 Supplier Medchemexpress LLC Supplier No. HY100795A5MG
Medchemexpress, HY-100795A 5mg Pirmenol (hydrochloride) CAS:61477-94-9 Pirmenol hydrochloride inhibits IK.ACh by blocking muscarinic receptors. The IC50 of Pirmenol for inhibition of Carbachol-induced IK.ACh is 0.1 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.