Learn More
Medchemexpress LLC Pirmenol hydrochloride | 61477-94-9 | MFCD01691662 | 98.8% | 374.95 g·mol⁻¹ | C22H31ClN2O | 100 MG
SDP
Catalog No. 5000212960
Encompass_Preferred
Pirmenol hydrochloride is an orally active antiarrhythmic research compound used in pharmacology and electrophysiology studies. It inhibits IK.ACh currents by blocking muscarinic acetylcholine receptors and is provided as a high-purity solid for laboratory research use.
  • Orally active compound suitable for cellular and in vivo studies.
  • Reported to inhibit IK.ACh with IC50 ≈ 0.1 μM.
  • High purity solid for reproducible experimental results.
  • Available in small-quantity packages appropriate for research workflows.
  • Intended for laboratory research; not for human or diagnostic use.
Catalog No. 50-002-12960 Supplier Medchemexpress LLC Supplier No. HY100795A100MG
Pirmenol hydrochloride is an orally active antiarrhythmic research compound used in pharmacology and electrophysiology studies. It inhibits IK.ACh currents by blocking muscarinic acetylcholine receptors and is provided as a high-purity solid for laboratory research use.
  • Orally active compound suitable for cellular and in vivo studies.
  • Reported to inhibit IK.ACh with IC50 ≈ 0.1 μM.
  • High purity solid for reproducible experimental results.
  • Available in small-quantity packages appropriate for research workflows.
  • Intended for laboratory research; not for human or diagnostic use.