Learn More
TARGETMOL CHEMICALS INC PIRMITEGRAVIR 5MG
SDP
Catalog No. 502580035
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Pirmitegravir(STP0404) is a potent and selective inhibitor of allosteric integrase (ALLINI) targeting the LEDGF/p75 binding site.Pirmitegravir inhibits PBMC. Pirmitegravir inhibits PBMC.Pirmitegravir has significant antiviral activity and may be useful in the study of HIV virus infection. purity: 99%
Catalog No. 50-258-0035 Supplier TARGETMOL CHEMICALS INC Supplier No. T397555MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Pirmitegravir(STP0404) is a potent and selective inhibitor of allosteric integrase (ALLINI) targeting the LEDGF/p75 binding site.Pirmitegravir inhibits PBMC. Pirmitegravir inhibits PBMC.Pirmitegravir has significant antiviral activity and may be useful in the study of HIV virus infection. purity: 99%