Learn More
AdipoGen Piroctone olamine
SDP
Catalog No. 502560241
Encompass_Preferred
Chemical. CAS 68890-66-4. Formula C14H23NO2 . C2H7NO. MW 237.3 . 61.1. Synthetic. Antimycotic compound with antidandruff efficacy. Selectively induced apoptosis in various human and murine myeloma and lymphoma cell lines. Compound can be used as analytical reference material.
Catalog No. 50-256-0241 Supplier AdipoGen Supplier No. CDXP0125M100
Chemical. CAS 68890-66-4. Formula C14H23NO2 . C2H7NO. MW 237.3 . 61.1. Synthetic. Antimycotic compound with antidandruff efficacy. Selectively induced apoptosis in various human and murine myeloma and lymphoma cell lines. Compound can be used as analytical reference material.