Learn More
Medchemexpress LLC Piroctone olamine | 68890-66-4 | 99.7% | 500 MG
SDP
Catalog No. 5000412074
Encompass_Preferred
Piroctone olamine is a pyridine derivative with a fungicidal effect. It acts as a hydroxypyridone anti-mycotic agent by penetrating cell membranes and forming complexes with iron ions, thereby inhibiting energy metabolism in mitochondria.
  • Fungicidal effect
  • Demonstrates low minimum inhibitory concentrations for Candida strains
  • Reduces fungal growth in intra-abdominal candidiasis in experimental models
Catalog No. 50-004-12074 Supplier Medchemexpress LLC Supplier No. HYB1345500MG
Piroctone olamine is a pyridine derivative with a fungicidal effect. It acts as a hydroxypyridone anti-mycotic agent by penetrating cell membranes and forming complexes with iron ions, thereby inhibiting energy metabolism in mitochondria.
  • Fungicidal effect
  • Demonstrates low minimum inhibitory concentrations for Candida strains
  • Reduces fungal growth in intra-abdominal candidiasis in experimental models