Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piroctone olamine, Thermo Scientific™

Catalog No. AC468820050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468820050 5 g
AC468820010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC468820050 Supplier Thermo Scientific Chemicals Supplier No. 468820050
  • $143.70
    $117.65 / Each of 1

    Save $26.05

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

Specifications

Alpha Vector PIROCTONEOLAMINE
Chemical Name or Material Piroctone olamine
CAS 68890-66-4
Melting Point 130°C to 138°C
Color White to Yellow
Infrared Spectrum Conforms
Packaging Glass bottle
Molecular Formula C14 H23 N O2 . C2 H7 N O
Quantity 5 g
MDL Number 01690792
Merck Index XIII 7585
Molecular Weight (g/mol) 298.46
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.