Learn More
Medchemexpress LLC Pironetin 5mg | 151519-02-7 | 5 MG
SDP
Catalog No. 5000227217
Encompass_Preferred
Pironetin is a research reagent that binds α-tubulin and inhibits microtubule polymerization, producing cell-cycle arrest and cytotoxic effects in various cell lines. It is supplied in small, measured milligram quantities for in vitro laboratory use.
  • Microtubule/tubulin inhibitor that binds α-tubulin.
  • Suitable for in vitro cell-cycle and cytotoxicity studies.
  • Available in small milligram pack sizes for assay development.
  • Chemical formula C19H32O4; molecular weight 324.45 g/mol.
  • Intended for research use only; not for human or diagnostic use.
Catalog No. 50-002-27217 Supplier Medchemexpress LLC Supplier No. HY1164465MG
Pironetin is a research reagent that binds α-tubulin and inhibits microtubule polymerization, producing cell-cycle arrest and cytotoxic effects in various cell lines. It is supplied in small, measured milligram quantities for in vitro laboratory use.
  • Microtubule/tubulin inhibitor that binds α-tubulin.
  • Suitable for in vitro cell-cycle and cytotoxicity studies.
  • Available in small milligram pack sizes for assay development.
  • Chemical formula C19H32O4; molecular weight 324.45 g/mol.
  • Intended for research use only; not for human or diagnostic use.