Learn More
Apexbio Technology LLC Piroxicam 36322-90-4 1g
SDP
Catalog No. 502240590
Encompass_Preferred
Piroxicam (CAS 36322-90-4) is a non-steroidal anti-inflammatory compound that acts as a dual inhibitor of cyclooxygenase isoforms COX-1 (human prostaglandin H synthase-1) and COX-2 (hPGHS-2) with greater potency toward COX-1 By suppressing prostaglandin synthesis piroxicam mediates anti-inflammatory effects and additionally blocks ADP-induced platelet aggregation This compound is utilized in research to study prostaglandin-mediated pathways evaluate cyclooxygenase inhibition profiles and investigate mechanisms underlying inflammation and platelet function
This item is not returnable. View return policy
Catalog No. 50-224-0590 Supplier Apexbio Technology LLC Supplier No. A85031000
Piroxicam (CAS 36322-90-4) is a non-steroidal anti-inflammatory compound that acts as a dual inhibitor of cyclooxygenase isoforms COX-1 (human prostaglandin H synthase-1) and COX-2 (hPGHS-2) with greater potency toward COX-1 By suppressing prostaglandin synthesis piroxicam mediates anti-inflammatory effects and additionally blocks ADP-induced platelet aggregation This compound is utilized in research to study prostaglandin-mediated pathways evaluate cyclooxygenase inhibition profiles and investigate mechanisms underlying inflammation and platelet function