Learn More
Selleck Chemical LLC Piroxicam 50mg 36322-90-4 CP 16171
SDP
Catalog No. 501361554
Encompass_Preferred
Piroxicam (CP 16171) is a non-selective COX inhibitor, used in the treatment of rheumatoid and osteoarthritis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1554 Supplier Selleck Chemical LLC Supplier No. S171350MG
Piroxicam (CP 16171) is a non-selective COX inhibitor, used in the treatment of rheumatoid and osteoarthritis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.