Learn More
Medchemexpress LLC Piroxicam-d3 | 942047-64-5 | 99.7% | 5 MG
SDP
Catalog No. 5000406686
Encompass_Preferred
Piroxicam-d3 is a deuterium-labeled version of Piroxicam, a non-steroidal anti-inflammatory drug (NSAID) and COX inhibitor. It is intended for research use only and can be utilized as a tracer and an internal standard for quantitative analysis.
  • Functions as a non-steroidal anti-inflammatory drug (NSAID)
  • Acts as a COX inhibitor with IC50 values of 47 μM for human monocyte COX-1 and 25 μM for human monocyte COX-2
  • Can be used as a tracer in research applications
  • Serves as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS
Catalog No. 50-004-06686 Supplier Medchemexpress LLC Supplier No. HYB0253S5MG
Piroxicam-d3 is a deuterium-labeled version of Piroxicam, a non-steroidal anti-inflammatory drug (NSAID) and COX inhibitor. It is intended for research use only and can be utilized as a tracer and an internal standard for quantitative analysis.
  • Functions as a non-steroidal anti-inflammatory drug (NSAID)
  • Acts as a COX inhibitor with IC50 values of 47 μM for human monocyte COX-1 and 25 μM for human monocyte COX-2
  • Can be used as a tracer in research applications
  • Serves as an internal standard for quantitative analysis using techniques like NMR, GC-MS, or LC-MS