Learn More
Medchemexpress LLC Piroxicam-d3 (CP-16171 d3) | 942047-64-5 | 99.7% | 334.36 g/mol | C15H10D3N3O4S | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB0253S10MG
Encompass_Preferred
Piroxicam-d3 is a deuterium-labeled analog of the nonsteroidal anti-inflammatory drug piroxicam. It is intended for research use as an internal standard or tracer in quantitative analytical methods such as LC-MS, GC-MS, and NMR; not for human or veterinary use.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High purity suitable for analytical workflows (≈99.7%).
  • Molecular weight 334.36 g/mol; formula C15H10D3N3O4S.
  • Supplied in small analytical quantities for method development and validation.
Catalog No. 50-002-98894