Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piroxicam, MP Biomedicals™

Catalog No. ICN15627701 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ICN15627701 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15627701 Supplier MP Biomedicals, Inc Supplier No. 0215627701

Specifications

CAS 36322-90-4
Color White
Molecular Formula C15H13N3O4S
Quantity 1 g
MDL Number MFCD00057317
Synonym piroxicam, feldene, piroxicamum, pyroxycam, roxicam, piroftal, baxo, artroxicam, bruxicam, caliment
InChI Key QYSPLQLAKJAUJT-UHFFFAOYSA-N
SMILES CN1C(=C(C2=CC=CC=C2S1(=O)=O)O)C(=O)NC3=CC=CC=N3
IUPAC Name 4-hydroxy-2-methyl-1,1-dioxo-N-pyridin-2-yl-1$l^{6},2-benzothiazine-3-carboxamide
Molecular Weight (g/mol) 331.346
PubChem CID 54676228
ChEBI CHEBI:8249
Formula Weight 331.35
Physical Form Solid
Chemical Name or Material Piroxicam
Show More Show Less
Recommended Storage Store at + 4°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.