Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piroxicam, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog No. 18605242 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-242 1 kg Amber Glass Bottle
18-605-241 100 g Amber Glass Bottle
18-605-243 5 kg Poly Pail
18-605-238 1 g Amber Glass Bottle
18-605-240 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-605-242 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P11981KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

P1198, 36322-90-4, C15H13N3O4S

Spectrum™ Chemical Piroxicam, USP is an anti-inflammatory drug of the oxicam class to treat painful and inflammatory conditions like arthritis. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 36322-90-4
Molecular Formula C15H13N3O4S
Molecular Weight (g/mol) 331.35
InChI Key LDJVNWFKNMYYNT-FYWRMAATSA-N
IUPAC Name (3E)-3-{hydroxy[(pyridin-2-yl)amino]methylidene}-2-methyl-3,4-dihydro-2H-1λ6,2-benzothiazine-1,1,4-trione
SMILES CN1\C(=C(\O)NC2=CC=CC=N2)C(=O)C2=CC=CC=C2S1(=O)=O

Specifications

CAS Max % 1
Ignition Residue 0.003
Packaging Amber Glass Bottle
Quantity 1 kg
Percent Purity 97 to 103%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.