Learn More
Cayman Chemical Pirtobrutinib, Purity: ≥98%, CAS: 2101700-15-4, 50 mg
SDP
Catalog No. 502910056
Encompass_Preferred
A BTK inhibitor (IC50s = 10-100 and C481S, respectively); selective for BTK over EGFR (IC50 = 1-10 µM); decreases the number of Ki67-positive cells in MEC-1 CLL mouse xenograft models using MEC-1 cells that express either wild-type BTK or BTKC481S
Catalog No. 50-291-0056 Supplier Cayman Chemical Supplier No. 3677550 MG
A BTK inhibitor (IC50s = 10-100 and C481S, respectively); selective for BTK over EGFR (IC50 = 1-10 µM); decreases the number of Ki67-positive cells in MEC-1 CLL mouse xenograft models using MEC-1 cells that express either wild-type BTK or BTKC481S