Learn More
Medchemexpress LLC HY-131328 5mg , Pirtobrutinib CAS:2101700-15-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1313285MG
Encompass_Preferred
HY-131328 5mg Pirtobrutinib CAS: 2101700-15-4 Pirtobrutinib (LOXO-305), a highly selective and non-covalent next generation BTK inhibitor, inhibits diverse BTK C481 substitution mutations. Pirtobrutinib causes regression of BTK-dependent lymphoma tumors in mouse xenograft models. Pirtobrutinib is also more than 300-fold selective for BTK versus 370 other kinases tested and shows no significant inhibition of non-kinase off-targets at 1 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-8981