Learn More
Selleck Chemical LLC Pirtobrutinib LOXO-305-5mg
SDP
Supplier:  Selleck Chemical LLC S98255MG
Encompass_Preferred
Pirtobrutinib (LOXO-305, LY 3527727, RXC-005) is a highly selective, non-covalent, next generation BTK inhibitor with an IC50 of 5.69 nM in WT BTK HEK cells. Pirtobrutinib shows more than 300-fold selective for BTK over 98% of 370 other kinases.
Catalog No. 50-238-9805