Learn More
CRYOBIOPHYSICA INC CRYOBIOPHYSICA INC PISEP BUFFER KIT 5L FOR WIDE RANGE PH GRADIENT CHROMATOGRAPHY
Catalog No. NC1469625
Encompass_Preferred
The pISep buffers allow you to create pH gradients over the pH range 2.4-10.8, and can also work perfectly well over any interval within that range. On a two channel HPLC or FPLC the software allows you to completely control single or multi-step, linear or nonlinear pH gradients over or within the range 2.4-10.8, even in the presence of isocratic levels of NaCl (0 to 1 M), urea (0 to 8 M), or acetonitrile (0 to 80% v/v). The software creates gradient instructions for any hplc or fplc. The buffers are compatible with most strong and weak anion and cation exchange stationary phases as well as mixed mode HIC HILIC and RP stationary phases. They are the most powerful way to create fully controlled pH gradients in chromatography.
Catalog No. NC1469625 Supplier CRYOBIOPHYSICA INC Supplier No. 20055
The pISep buffers allow you to create pH gradients over the pH range 2.4-10.8, and can also work perfectly well over any interval within that range. On a two channel HPLC or FPLC the software allows you to completely control single or multi-step, linear or nonlinear pH gradients over or within the range 2.4-10.8, even in the presence of isocratic levels of NaCl (0 to 1 M), urea (0 to 8 M), or acetonitrile (0 to 80% v/v). The software creates gradient instructions for any hplc or fplc. The buffers are compatible with most strong and weak anion and cation exchange stationary phases as well as mixed mode HIC HILIC and RP stationary phases. They are the most powerful way to create fully controlled pH gradients in chromatography.