Learn More
StatLab PiSmart ACS Full Tray
SDP
Catalog No. 22000198
Change view
Click to view available options
Includes:
1 Full Tray
Catalog No. Includes
22-000-198 1 Full Tray
1 options
Catalog No. 22-000-198 Supplier StatLab Supplier No. JAP210200A
Add to Cart
Add to Cart
Automatically collects and organizes up to 130 printed cassettes for efficient handling.
  • Supports High-volume, batch cassette printing workflows
  • Automatically collects and organizes up to 130 printed cassettes
  • Helps maintain an organized and efficient cassette handling process
  • Designed for rapid installation and easy operation
  • Supports a clean, controlled, and uninterrupted workflow
  • Includes one tray
Specifications
Product Type Collection Tray
Includes 1 Full Tray