Learn More
StatLab PiSmart ACS Small Tray Set
Description
- Automatically collects printed cassettes for efficient batch processing.
- Accommodates and organizes up to 130 printed cassettes.
- Supports laboratories with demanding batch cassette printing requirements.
- Keeps printed cassettes orderly for easier handling and retrieval.
- Simple to use and integrates smoothly into routine workflows.
- Reduces manual handling and helps maintain continuous cassette printing.
- Promotes an organized and controlled laboratory workspace.
- Set includes two smaller trays that hold 40 cassettes each and a larger tray that holds 50 cassettes.
Specifications
Specifications
| Product Type | Collection Tray |
| Includes | 3 Tray Set |