Learn More
StatLab PiSmart Biopsy Laser Cassettes
SDP
Catalog No. 21100728
Change view
Click to view available options
Color:
Green
Pink
Tan
White
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-100-728 Green
21-100-722 White
21-100-723 Tan
21-100-724 Pink
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 21-100-728 Supplier StatLab Supplier No. CASLPI100BXGR
Add to Cart
Add to Cart
This item is not returnable. View return policy
Biopsy laser cassettes, threaded with lid.
  • 1,000 per box, 2 box per case, 2,000 per case
  • Threaded in strips of 40 with lids separate
  • PiSmart Cassettes contain 4 patented pips at all 4 corners on the front of each cassette, protecting the thermal printing from high heat during dewaxing
  • Engineered specifically for use in the PiSmart Laser Cassette printer
  • Available in 9 colors
Specifications
Color Green
Product Type Biopsy Cassette