Learn More
StatLab PiSmart CL1 Single Hopper Laser Cassette Printer
Description
- Designed for on-demand printing, the PiSmart CL1 Single Hopper Laser Cassette
- Printer can deliver the first printed cassette in as little as 3 seconds
- Superior print quality produces durable print with excellent contrast and supports up to 200 characters, 600 dpi
- Unit has 1 Hopper with the ability to easily swap out hoppers for additional colors, as well as bypass the hopper and manually drop in a different type of cassette
- Integrated label designer, no additional software needed
- Small footprint is ideal for on-demand printing where space is limited
- High resolution touch screen monitor with an intuitive menu structure makes the printer easy to learn and use
- Fully Integrated barcode scanner supports the ability to scan data from a request form or tissue sample barcode
- User login and tracking creates traceability for root cause analysis
- Intelligent cassette selection prints the right type and number of cassettes based on the protocol selected for each tissue type
- Kit includes 4 years and 1 year factory warranty of StatSwap service coverage, Lifetime printhead guarantee, Guaranteed LIS compatibility, Guaranteed validated consumables (cassettes), PathSmart tracking system hub module the PiSmart promise
Specifications
Specifications
| Product Type | PiSmart CL1 Printer |
| For Use With (Equipment) | PathSmart Tracking System |