Learn More
StatLab PiSmart CL1 Single Hopper Laser Cassette Printer
SDP
Catalog No. 21010280
Change view
Click to view available options
Product Type:
PiSmart CL1 Printer
PiSmart CL1 Printer Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
21-010-280 PiSmart CL1 Printer Kit
21-010-279 PiSmart CL1 Printer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-010-280 Supplier StatLab Supplier No. PISMARTCL1HOPPERPSPKIT
Add to Cart
Add to Cart
High resolution touch screen monitor with an intuitive menu structure makes the printer easy to learn and use
  • Designed for on-demand printing, the PiSmart CL1 Single Hopper Laser Cassette
  • Printer can deliver the first printed cassette in as little as 3 seconds
  • Superior print quality produces durable print with excellent contrast and supports up to 200 characters, 600 dpi
  • Unit has 1 Hopper with the ability to easily swap out hoppers for additional colors, as well as bypass the hopper and manually drop in a different type of cassette
  • Integrated label designer, no additional software needed
  • Small footprint is ideal for on-demand printing where space is limited
  • High resolution touch screen monitor with an intuitive menu structure makes the printer easy to learn and use
  • Fully Integrated barcode scanner supports the ability to scan data from a request form or tissue sample barcode
  • User login and tracking creates traceability for root cause analysis
  • Intelligent cassette selection prints the right type and number of cassettes based on the protocol selected for each tissue type
  • Kit includes 4 years and 1 year factory warranty of StatSwap service coverage, Lifetime printhead guarantee, Guaranteed LIS compatibility, Guaranteed validated consumables (cassettes), PathSmart tracking system hub module the PiSmart promise
Specifications
Product Type PiSmart CL1 Printer Kit
For Use With (Equipment) PiSmart S1 Printer