Learn More
StatLab PiSmart Hopper Thermal Transfer Cassette Printer
Description
- Designed for on-demand printing, the PiSmart Single Plus can deliver the first printed cassette in as little as 3 seconds
- Superior print quality produces dark, durable print with excellent contrast and supports up to 200 characters 300 dpi
- Fully Integrated barcode scanner supports the ability to scan data from a request form or tissue sample barcode
- PathSmart Innovative Tracking Solution connects printers to a hub and delivers reporting data to help close gaps in chain of custody
- Intelligent cassette selection prints the right type and number of cassettes based on the protocol selected for each tissue type
- Easy LIS Integration and Flexible Connectivity - connects to any available on-market LIS
- 1-year factory warranty
Specifications
Specifications
| Type | Thermal Transfer Cassette Printer |
| Voltage | 120/240 V |
| Color | Gray |