Learn More
StatLab PiSmart MicroBiopsy Laser Cassettes
SDP
Catalog No. 21100712
Change view
Click to view available options
Color:
Gray
Lavender
Pink
Yellow
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-100-712 Yellow
21-100-715 Pink
21-100-717 Lavender
21-100-718 Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 21-100-712 Supplier StatLab Supplier No. CASLPI100MBXYL
Add to Cart
Add to Cart
This item is not returnable. View return policy
Microbiopsy laser cassettes, threaded with lid.
  • 1,000 per box, 2 box per case, 2,000 per case
  • Threaded in strips of 40 with lids separate
  • PiSmart Cassettes contain 4 patented pips at all 4 corners on the front of each cassette, protecting the thermal printing from high heat during dewaxing
  • Engineered specifically for use in the PiSmart Laser Cassette printer
  • Available in 9 colors
Specifications
Color Yellow
Product Type MicroBiopsy Cassette