Learn More
StatLab PiSmart S1 Single Hopper Thermal Transfer Slide Printer - Trade-In
SDP
Catalog No. 21402112
Change view
Click to view available options
Closure Included:
Yes
Catalog No. Closure Included
21-402-112 Yes
1 options
Catalog No. 21-402-112 Supplier StatLab Supplier No. PISMARTS1PSPKITTRADE
Add to Cart
Add to Cart
Thermal transfer slide printer includes 1-year factory warranty and PathSmart tracking system hub - trade-in of old printer required
  • Designed for on-demand printing, the PiSmart S1 Single Hopper Thermal Transfer Slide Printer can deliver the first printed slide in as little as 3 seconds.
  • Superior print quality produces dark, durable print with excellent contrast and supports up to 200 characters: 300 dpi.
  • Users have the ability to easily swap hoppers, as well as bypass the hopper and manually drop in a different type of slide.
  • Integrated label designer: no additional software needed.
  • Small footprint is ideal for on-demand printing where space is limited.
  • High resolution touch screen monitor with an intuitive menu structure makes the printer easy to learn and use.
  • Built-in tools for accuracy and workflow automation.
  • Fully integrated barcode scanner supports the ability to scan data from a request form or tissue sample barcode.
  • PathSmart Innovative Tracking Solution connects printers to a hub and delivers reporting data to help close gaps in chain of custody.
  • User login and tracking creates traceability for root cause analysis.
  • Intelligent slide selection prints the right type and number of slides based on the protocol selected for each tissue type.
  • Easy LIS integration and flexible connectivity—connects to any available on-market LIS.
  • Printer includes 1-year factory warranty.
  • Trade-In of old printer required.
Specifications
Product Type PiSmart S1 Printer
Closure Included Yes