Learn More
StatLab™ PiSmart Six 6-Hopper Thermal Transfer Cassette Printer Kit
Description
- Designed for on-demand printing, the PiSmart Six can deliver the first printed cassette in as little as 3 seconds.
- Superior print quality produces dark, durable print with excellent contrast and supports up to 200 characters: 300 dpi.
- Unit has 6 hoppers with the ability to easily swap out hoppers for additional colors, as well as bypass the 6 hoppers and manually drop in a different type of cassette.
- Integrated label designer; no additional software needed.
- Small footprint is ideal for on-demand printing where space is limited.
- High resolution touch screen monitor with an intuitive menu structure makes the printer easy to learn and use.
- Built-in tools for accuracy and workflow automation.
- Fully integrated barcode scanner supports the ability to scan data from a request form or tissue sample barcode.
- PathSmart Innovative Tracking Solution connects printers to a hub and delivers reporting data to help close gaps in chain of custody.
- User login and tracking creates traceability for root cause analysis.
- Intelligent cassette selection prints the right type and number of cassettes based on the protocol selected for each tissue type.
- Easy LIS Integration and Flexible Connectivity - connects to any available on-market LIS.
- Validated PiSmart cassettes for use with the PiSmart Cassette Printer and manufactured by StatLab™, the proprietary design of PiSmart cassettes ensures print integrity is retained through the entire workflow.
- Kit includes printer, 4 years of StatSwap service coverage, lifetime printhead guarantee, guaranteed LIS compatibility, guaranteed validated consumables (cassettes), PathSmart tracking system hub module, and 10 thermal transfer ribbons.
Specifications
Specifications
| Type | Thermal Transfer Cassette Printer |
| Voltage | 110/240 V |
| Color | White |
| Product Type | Printer Kit |