Learn More
StatLab PiSmart Six Hopper Laser Cassette Printer
SDP
Catalog No. PISMARTCL6
Change view
Click to view available options
Color:
Gray
Catalog No. Color
PISMARTCL6 Gray
1 options
Catalog No. PISMARTCL6 Supplier StatLab Supplier No. PISMARTCL6
Add to Cart
Add to Cart
Includes a 1-year factory warranty and PathSmart tracking system hub
  • Designed for on-demand printing, the PiSmart Six Hopper Laser Cassette Printer can deliver the first printed cassette in as little as 3 seconds
  • Superior print quality produces durable print with excellent contrast and supports up to 200 characters, 600 dpi
  • Unit has 6 Hoppers with the ability to easily swap out hoppers for additional colors, as well as bypass the 6 hoppers and manually drop in a different type of cassette
  • Fully Integrated barcode scanner supports the ability to scan data from a request form or tissue sample barcode
  • PathSmart innovative tracking solution connects printers to a hub and delivers reporting data to help close gaps in chain of custody
  • Easy LIS Integration and flexible connectivity - connects to any available on-market LIS
  • 1-year factory warranty
Specifications
Type Laser Printer
Voltage 120/240 V
Color Gray