Learn More
StatLab PiSmart Six Hopper Laser Cassette Printer Kit
SDP
Catalog No. 21100702
Change view
Click to view available options
Color:
White
Catalog No. Color
21-100-702 White
1 options
110/240V power adapter 24V DC
Catalog No. 21-100-702 Supplier StatLab Supplier No. PISMARTCL6HOPPERPSPKIT
Add to Cart
Add to Cart
110/240V power adapter 24V DC
Printer Kit includes: printer, 4 years of StatSwap service coverage, lifetime printhead guarantee, guaranteed LIS compatibility, guaranteed validated consumables (cassettes), and PathSmart tracking system hub module.
  • Designed for on-demand printing, the PiSmart Six Hopper Laser Cassette Printer can deliver the first printed cassette in as little as 3 seconds
  • Superior print quality produces durable print with excellent contrast and supports up to 200 characters, 600 dpi
  • Unit has 6 Hoppers with the ability to easily swap out hoppers for additional colors, as well as bypass the 6 hoppers and manually drop in a different type of cassette
  • Integrated label designer, no additional software needed
  • Small footprint is ideal for on-demand printing where space is limited
  • High resolution touch screen monitor with an intuitive menu structure makes the printer easy to learn and use
  • Built-in tools for accuracy and workflow automation
  • Fully Integrated barcode scanner supports the ability to scan data from a request form or tissue sample barcode
  • PathSmart innovative tracking solution connects printers to a hub and delivers reporting data to help close gaps in chain of custody
  • User login and tracking creates traceability for root cause analysis
  • Intelligent cassette selection prints the right type and number of cassettes based on the protocol selected for each tissue type
  • Easy LIS Integration and flexible connectivity - connects to any available on-market LIS
  • Validated PiSmart Laser Cassettes for use with the PiSmart Laser Cassette Printer and manufactured by StatLab; the proprietary design of PiSmart Cassettes ensures print integrity is retained through the entire workflow
  • Kit includes:
    • Printer
    • 4 years of StatSwap service coverage
    • Lifetime printhead guarantee
    • Guaranteed LIS compatibility
    • Guaranteed validated consumables (cassettes)
    • PathSmart tracking system hub module - the PiSmart promise
Specifications
Type Laser Printer
Voltage 110/240 V
Color White
4-Years of StatSwap Service requires the use of PiSmart Validated Cassettes on this printer.