Learn More
StatLab PiSmart Slide Printer Hopper
SDP
Catalog No. 21124400
Change view
Click to view available options
Product Type:
Slide Printer
Catalog No. Product Type
21-124-400 Slide Printer
1 options
Catalog No. 21-124-400 Supplier StatLab Supplier No. PISMARTS2HOPPER
Add to Cart
Add to Cart
PiSmart Slide Printer Hopper 72-slide capacity
  • Use with PiSmart Slide Printers with standard slides
  • Each hopper can hold 72 slides
  • Keep extra hoppers on hand already loaded with a variety of slides to easily switch between different slides
Specifications
Product Type Slide Printer
Quantity 1 Ea.