Learn More
StatLab PiSmart Slotted Laser Embedding Cassettes
SDP
Catalog No. 21100707
Change view
Click to view available options
Color:
Blue
Gray
Green
Lavender
Orange
Pink
Tan
White
Yellow
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
21-100-707 Orange
21-100-711 Blue
21-100-703 Yellow
21-100-704 White
21-100-705 Tan
21-100-706 Pink
21-100-708 Lavender
21-100-709 Gray
21-100-710 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. 21-100-707 Supplier StatLab Supplier No. CASLPI100SLOR
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Slotted laser embedding cassettes, threaded with lid.
  • 1,000 per box, 2 box per case, 2,000 per case
  • Threaded in strips of 40 with lids separate
  • PiSmart Cassettes contain 4 patented pips at all 4 corners on the front of each cassette, protecting the thermal printing from high heat during dewaxing
  • Engineered specifically for use in the PiSmart Laser Cassette printer
  • Available in 9 colors
Specifications
Color Orange
Product Type Slotted Cassette