Learn More
StatLab™ PiSmart Slotted Thermal Transfer Embedding Cassettes
SDP
Catalog No. 22235692
Click to view available options
Color:
Blue
Gray
Green
Lavender
Orange
Pink
Tan
White
Yellow
This item is not returnable. View return policy
This item is not returnable. View return policy
Threaded with lid.
  • 1,000 per box, with 2 boxes per case for a total of 2,000 per case.
  • Threaded in strips of 40 with lids separate.
  • PiSmart cassettes contain 4 patented pips at all 4 corners on the front of each cassette, protecting the thermal printing from high heat during dewaxing.
  • Engineered specifically for use in the PiSmart Thermal Transfer Cassette printer.
  • Available in 9 colors.
Specifications
Color Lavender
Product Type Embedding Cassette
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.