Learn More
StatLab™ PiSmart Slotted Thermal Transfer Embedding Cassettes
SDP
Catalog No. p-200049723
Change view
Click to view available options
Color:
Blue
Gray
Green
Pink
White
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-235-694 Yellow
22-235-695 Pink
22-243-553 Blue
22-244-027 Gray
22-244-028 Green
22-244-029 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 22-235-694 Supplier StatLab™ Supplier No. EAM020972B
This item is not returnable. View return policy
Threaded with lid.
  • 1,000 per box, with 2 boxes per case for a total of 2,000 per case.
  • Threaded in strips of 40 with lids separate.
  • PiSmart cassettes contain 4 patented pips at all 4 corners on the front of each cassette, protecting the thermal printing from high heat during dewaxing.
  • Engineered specifically for use in the PiSmart Thermal Transfer Cassette printer.
  • Available in 9 colors.
Specifications
Color Yellow
Product Type Embedding Cassette