Learn More
StatLab™ PiSmart Thermal Transfer Microbiopsy Cassettes
SDP
Catalog No. 22235682
Change view
Click to view available options
Color:
Blue
Green
Lavender
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-235-682 White
22-235-674 Lavender
22-235-676 Yellow
22-235-679 Blue
22-235-681 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 22-235-682 Supplier StatLab™ Supplier No. EAM040272B
This item is not returnable. View return policy
Threaded with lid.
  • 1,000 per box, with 2 boxes per case for a total of 2,000 per case.
  • Threaded in strips of 40 with lids separate.
  • PiSmart cassettes contain 4 patented pips at all 4 corners on the front of each cassette, protecting the thermal printing from high heat during dewaxing.
  • Engineered specifically for use in the PiSmart Thermal Transfer Cassette printer.
  • Available in 9 colors.
Specifications
Color White
Product Type Microbiopsy Cassette