Learn More
StatLab™ PiSmart Thermal Transfer Microbiopsy Cassettes
SDP
Catalog No. p-200049727
Change view
Click to view available options
Color:
Blue
Green
Lavender
White
Yellow
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
22-235-674 Lavender
22-235-676 Yellow
22-235-679 Blue
22-235-681 Green
22-235-682 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 22-235-674 Supplier StatLab™ Supplier No. EAM041972B
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Threaded with lid.
  • 1,000 per box, with 2 boxes per case for a total of 2,000 per case.
  • Threaded in strips of 40 with lids separate.
  • PiSmart cassettes contain 4 patented pips at all 4 corners on the front of each cassette, protecting the thermal printing from high heat during dewaxing.
  • Engineered specifically for use in the PiSmart Thermal Transfer Cassette printer.
  • Available in 9 colors.
Specifications
Color Lavender
Product Type Microbiopsy Cassette