Learn More
StatLab PiSmart Twin-Hopper Thermal Slide Printer Kit
SDP
Catalog No. 21124401
Change view
Click to view available options
Product Type:
Slide Printer
Catalog No. Product Type
21-124-401 Slide Printer
1 options
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Catalog No. 21-124-401 Supplier StatLab Supplier No. PISMARTS2PSPKIT
Add to Cart
Add to Cart
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Avoid risks associated with batch printing by using the small, efficient on-demand PiSmart Slide Printer.
  • Designed for on-demand printing, the PiSmart Twin Hopper Slide Printer can deliver the first printed slide in as little as 3 seconds.
  • Superior print quality produces dark, durable print with excellent contrast and supports up to 200 characters: 300 dpi.
  • Users have the ability to easily swap hoppers, as well as bypass the hoppers and manually drop in a different type of slide.
  • Integrated label designer: no additional software needed.
  • Small footprint is ideal for on-demand printing where space is limited.
  • High resolution touch screen monitor with an intuitive menu structure makes the printer easy to learn and use.
  • Built-in tools for accuracy and workflow automation.
  • Fully integrated barcode scanner supports the ability to scan data from a request form or tissue sample barcode.
  • PathSmart Innovative Tracking Solution connects printers to a hub and delivers reporting data to help close gaps in chain of custody.
  • User login and tracking creates traceability for root cause analysis.
  • Intelligent slide selection prints the right type and number of slides based on the protocol selected for each tissue type.
  • Easy LIS integration and flexible connectivity - connects to any available on-market LIS.
  • Kit includes printer, 4 years of StatSwap service coverage, lifetime printhead guarantee, guaranteed LIS compatibility, guaranteed validated consumables (slides), PathSmart tracking system hub module, and 10 thermal transfer ribbons.
Specifications
Product Type Slide Printer
Quantity 1 Kit