Learn More
Waters Corp Piston Assembly 700000161
Supplier:  Waters Corp 700000161
Encompass_Preferred
Made from the highest quality sapphire, this plunger preserves the life span of the plunger seal. Precise roundness and smoothness tolerances minimize the wear of the plunger seal. Careful concentric alignment of the plunger within its stainless steel holder ensures that the sapphire rides directly into the center of the plunger seal, reducing wear of both parts.
Catalog No. 50-792-320