Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Piston Pump - Electro Zinc Plated Steel

Catalog No. 18666732 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
30 and 55 Gallon Drums
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
18-666-732 30 and 55 Gallon Drums
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-666-732 Supplier Justrite™ Supplier No. 07712
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ideal for small quantity dispensing of liquids. This manually operated piston pump is FM approved, ideal for transferring flammable and combustible liquids in a controlled and safe manner.

  • Ideal for small quantity dispensing of liquids from 30 and 55 gallon drums.
  • This manually operated piston pump is FM approved and delivers up to 8 oz. per stroke, ideal for transferring flammable and combustible liquids in a controlled and safe manner.
  • The pump body is constructed of Electro Zinc plated steel with PTFE seals, offering superior chemical resistance and a long life.
  • Included with the pump is a standard two-inch NPT threaded bung adapter.
  • The included pick-up tube allows the pump to reach up to 36 inches below the lid of the drum.
  • Included flash arrestor keeps contents safe from external flame flashbacks.
  • The bonding and grounding clamps come pre-assembled and provide the means to safely ground the receiving container to avoid a static discharge.

Specifications

Capacity (English) 8.0 oz./Stroke
Capacity (Metric) 0.24 L/Stroke
Color Silver
For Use With (Application) Dispensing of Flammable Liquids
For Use With (Equipment) 30 and 55 Gallon Drums
Material Zinc-plated Steel
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.