Learn More
Justrite™ Piston Pump - Stainless Steel
Catalog No. 18666734 Shop All Justrite Safety Group Products
Change view
Click to view available options
For Use With (Equipment):
30 and 55 Gallon Drums
5 Gallon Pails
Capacity (English):
3.5 oz./Stroke
8.0 oz./Stroke
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Capacity (English)
18-666-734 30 and 55 Gallon Drums 8.0 oz./Stroke
18-666-733 5 Gallon Pails 3.5 oz./Stroke
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-666-734 Supplier Justrite™ Supplier No. 07717
Add to Cart
Add to Cart
Ideal for small quantity dispensing of liquids. This manually operated piston pump is FM approved, ideal for transferring flammable and combustible liquids in a controlled and safe manner.
  • Ideal for small quantity dispensing of liquids
  • This manually operated piston pump is FM approved, ideal for transferring flammable and combustible liquids in a controlled and safe manner.
  • The pump body is constructed of stainless steel with PTFE seals, offering superior chemical resistance and a long life.
  • Included flash arrestor keeps contents safe from external flame flashbacks.
  • The bonding and grounding clamps come pre-assembled and provide the means to safely ground the receiving container to avoid a static discharge.
Specifications
Capacity (English) 8.0 oz./Stroke
Capacity (Metric) 0.24 L/Stroke
For Use With (Application) Dispensing of Flammable Liquids
For Use With (Equipment) 30 and 55 Gallon Drums
Material Stainless Steel
Includes Standard two-inch NPT threaded bung adapter
Sterility Non-sterile