Learn More
Eldex Laboratories Inc PISTON SEAL ASSEMBLY KIT
Catalog No. NC1907326
Encompass_Preferred
Piston seal assembly kit; includes one graphite fiber reinforced PTFE seal, and tools for installation
Catalog No. NC1907326 Supplier Eldex Laboratories Inc Supplier No. 5941
Piston seal assembly kit; includes one graphite fiber reinforced PTFE seal, and tools for installation