Learn More
Analytical Sales & Services Piston Seals for Agilent 1050, 1100, 1200, 1260
Catalog No. 5000804252
Encompass
Agilent 1050 1100 1200 1260 1290 Analytical Pumps Replacement UHMWPE Piston Seals
Qty/Pkg: 10PK
Catalog No. 50-008-04252 Supplier Analytical Sales & Services Supplier No. 920055
Add to Cart
Add to Cart
Agilent 1050 1100 1200 1260 1290 Analytical Pumps Replacement UHMWPE Piston Seals
Qty/Pkg: 10PK