Learn More
Bioworld Pisum sativum (Pea) Lectin (PSA/PEA) - Texas Red
Catalog No. 50199159
Encompass
Technical Specifications:Ligand Source:Pisum sativumMolecular Weight:53 kDaCarbohydrate Specificity:αMan, Fucα6GlcNAcInhibiting/Eluting Sugar:a-Methylglucoside +a-MethylmannosideCarbohydrate for Elution:0.2Ma-Methylglucoside +a-MethylmannosideBlood Group Specificity:N/AConjugate: Texas RedForm:LiquidIsoelectric (pI):6.0-6.7Storage: -20°CApplication note: Calcium and Manganese are required for binding.
Catalog No. 50-199-159 Supplier Bioworld Supplier No. 217610471
Technical Specifications:Ligand Source:Pisum sativumMolecular Weight:53 kDaCarbohydrate Specificity:αMan, Fucα6GlcNAcInhibiting/Eluting Sugar:a-Methylglucoside +a-MethylmannosideCarbohydrate for Elution:0.2Ma-Methylglucoside +a-MethylmannosideBlood Group Specificity:N/AConjugate: Texas RedForm:LiquidIsoelectric (pI):6.0-6.7Storage: -20°CApplication note: Calcium and Manganese are required for binding.