Learn More
Bioworld Pisum sativum (Pea) Lectin (PSA/PEA) - FITC
Catalog No. 50199157
Encompass
FITC Conjugated Pisum sativum Lectin (Garden Pea) -PEA, PSA-Technical Specifications:Ligand Source:Pisum sativumMolecular Weight: 53 kDaCarbohydrate Specificity:αMan, Fucα6GlcNAcInhibiting/Eluting Sugar:a-Methylglucoside +a-MethylmannosideCarbohydrate for Elution: 0.2Ma-Methylglucoside +a-MethylmannosideBlood Group Specificity: N/AConjugate: FITCForm: LiquidIsoelectric (pI): 6.0-6.7Storage: -20°CApplication note: Calcium and Manganese are required for binding.
Catalog No. 50-199-157 Supplier Bioworld Supplier No. 217610451
FITC Conjugated Pisum sativum Lectin (Garden Pea) -PEA, PSA-Technical Specifications:Ligand Source:Pisum sativumMolecular Weight: 53 kDaCarbohydrate Specificity:αMan, Fucα6GlcNAcInhibiting/Eluting Sugar:a-Methylglucoside +a-MethylmannosideCarbohydrate for Elution: 0.2Ma-Methylglucoside +a-MethylmannosideBlood Group Specificity: N/AConjugate: FITCForm: LiquidIsoelectric (pI): 6.0-6.7Storage: -20°CApplication note: Calcium and Manganese are required for binding.