Learn More
Bioworld Pisum sativum (Pea) Lectin (PSA/PEA) - Rhodamine (TRITC)
Catalog No. 50199158
Encompass
Technical Specifications:Ligand Source:Pisum sativumMolecular Weight:53 kDaCarbohydrate Specificity:αMan, Fucα6GlcNAcInhibiting/Eluting Sugar:a-Methylglucoside +a-MethylmannosideCarbohydrate for Elution:0.2Ma-Methylglucoside +a-MethylmannosideBlood Group Specificity:N/AConjugate: RhodamineForm:LiquidIsoelectric (pI):6.0-6.7Storage: -20°CApplication note: Calcium and Manganese are required for binding.
Catalog No. 50-199-158 Supplier Bioworld Supplier No. 217610461
Technical Specifications:Ligand Source:Pisum sativumMolecular Weight:53 kDaCarbohydrate Specificity:αMan, Fucα6GlcNAcInhibiting/Eluting Sugar:a-Methylglucoside +a-MethylmannosideCarbohydrate for Elution:0.2Ma-Methylglucoside +a-MethylmannosideBlood Group Specificity:N/AConjugate: RhodamineForm:LiquidIsoelectric (pI):6.0-6.7Storage: -20°CApplication note: Calcium and Manganese are required for binding.