Learn More
TARGETMOL CHEMICALS INC PITAVASTATIN CALCIUM 100MG
SDP
Catalog No. 502580759
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pitavastatin calcium (NK-104) is a potent inhibitor of HMG-CoA reductase (Ki 1.7 nM). It lowers both total cholesterol and low-density lipoprotein cholesterol in animals and humans. Metabolism of pitavastatin by the cytochrome P450 system is minimal reducing the risk of drug-drug interactions. purity: 99%
Catalog No. 50-258-0759 Supplier TARGETMOL CHEMICALS INC Supplier No. T2534100MG
Also available in 5 mg 10 mg 25 mg 50 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Pitavastatin calcium (NK-104) is a potent inhibitor of HMG-CoA reductase (Ki 1.7 nM). It lowers both total cholesterol and low-density lipoprotein cholesterol in animals and humans. Metabolism of pitavastatin by the cytochrome P450 system is minimal reducing the risk of drug-drug interactions. purity: 99%