Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pitavastatin Calcium | 147526-32-7 | 99.89% | 1 ML
SDP

Catalog No. 5000450920
Encompass_Preferred

Pitavastatin Calcium | 147526-32-7 | 99.89% | 1 ML

Catalog No. 50-004-50920 Supplier Medchemexpress LLC Supplier No. HYB014410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pitavastatin Calcium | 147526-32-7 | 99.89% | 1 ML

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.