Learn More
Selleck Chemical LLC Pitavastatin calcium S1759-1g
SDP
Catalog No. 503109134
Encompass_Preferred
Pitavastatin calcium a novel member of the medication class of statins is a calcium salt formulation of pitavastatin which is a highly effective HMG-CoA reductase inhibitor Pitavastatin Calcium attenuates AGEs-induced mitophagy via inhibition of ROS generation Pitavastatin Calcium induces autophagy and apoptosis
Catalog No. 50-310-9134 Supplier Selleck Chemical LLC Supplier No. S17591G
Pitavastatin calcium a novel member of the medication class of statins is a calcium salt formulation of pitavastatin which is a highly effective HMG-CoA reductase inhibitor Pitavastatin Calcium attenuates AGEs-induced mitophagy via inhibition of ROS generation Pitavastatin Calcium induces autophagy and apoptosis