Learn More
Selleck Chemical LLC Pitavastatin calcium S1759-200mg
SDP
Catalog No. 503110924
Encompass_Preferred
Pitavastatin calcium a novel member of the medication class of statins is a calcium salt formulation of pitavastatin which is a highly effective HMG-CoA reductase inhibitor Pitavastatin Calcium attenuates AGEs-induced mitophagy via inhibition of ROS generation Pitavastatin Calcium induces autophagy and apoptosis
Catalog No. 50-311-0924 Supplier Selleck Chemical LLC Supplier No. S1759200MG
Pitavastatin calcium a novel member of the medication class of statins is a calcium salt formulation of pitavastatin which is a highly effective HMG-CoA reductase inhibitor Pitavastatin Calcium attenuates AGEs-induced mitophagy via inhibition of ROS generation Pitavastatin Calcium induces autophagy and apoptosis