Learn More
Selleck Chemical LLC Pitavastatin Calcium 50mg 147526-32-7 NK-104, P-872441, itavastatin, nisvastatin
SDP
Catalog No. 505398
Encompass_Preferred
Pitavastatin Calcium (NK-104, P-872441, itavastatin, nisvastatin), a novel member of the medication class of statins, is a calcium salt formulation of pitavastatin which is a highly effective HMG-CoA reductase inhibitor. Pitavastatin Calcium attenuates AGEs-induced mitophagy via inhibition of ROS generation. Pitavastatin Calcium induces autophagy and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-539-8 Supplier Selleck Chemical LLC Supplier No. S175950MG
Pitavastatin Calcium (NK-104, P-872441, itavastatin, nisvastatin), a novel member of the medication class of statins, is a calcium salt formulation of pitavastatin which is a highly effective HMG-CoA reductase inhibitor. Pitavastatin Calcium attenuates AGEs-induced mitophagy via inhibition of ROS generation. Pitavastatin Calcium induces autophagy and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.