Learn More
Medchemexpress LLC Pitavastatin-d4-1 sodium | 147511-69-1 | 447.47 | C25H19D4FNNaO4 | 10 MG

Supplier: Medchemexpress LLC HYB0144AS310MG
Pitavastatin-d4-1 sodium is a deuterium-labelled analogue of pitavastatin sodium intended as an internal standard and tracer for quantitative analytical methods such as LC-MS, GC-MS, and NMR. Supplied as a solid for research use only, it supports method development and accurate quantitation in biological and chemical analyses.
- Deuterium-labeled internal standard for quantitative analysis.
- Suitable for LC-MS, GC-MS, and NMR applications.
- Solid form, supplied as a 10 MG package.
- Molecular weight 447.47 and chemical formula C25H19D4FNNaO4.
- Recommended storage: sealed, away from moisture; in solvent -80°C (up to 6 months) or -20°C (up to 1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.