Learn More
Medchemexpress LLC Pitavastatin-d4-1 (sodium) | 2070009-71-9 | >98% | 1 MG
SDP
Catalog No. 5000450903
Encompass_Preferred
Pitavastatin D4 is a deuterium-labeled form of Pitavastatin, a potent HMG-CoA reductase inhibitor. This compound is suitable for use as a laboratory chemical in research and substance manufacturing.
  • Deuterium labeled compound
  • Functions as a potent HMG-CoA reductase inhibitor
  • Ideal for laboratory research applications
  • Purity greater than 98%
Catalog No. 50-004-50903 Supplier Medchemexpress LLC Supplier No. HYB0144AS1MG
Pitavastatin D4 is a deuterium-labeled form of Pitavastatin, a potent HMG-CoA reductase inhibitor. This compound is suitable for use as a laboratory chemical in research and substance manufacturing.
  • Deuterium labeled compound
  • Functions as a potent HMG-CoA reductase inhibitor
  • Ideal for laboratory research applications
  • Purity greater than 98%